Assay Details
| Gene Symbol: 9530068E07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153117.2 | NP_694757.1 | 2 - 3 | 695 | 74 | |
| GenBank mRNA | AK007314.1 | - | 2 - 3 | 557 | 74 | |
| AK079269.1 | - | 2 - 3 | 696 | 74 | ||
| AK135934.1 | - | 2 - 3 | 659 | 74 | ||
| AK144677.1 | - | 2 - 3 | 659 | 74 | ||
| AK145254.1 | - | 2 - 3 | 664 | 74 | ||
| AK147956.1 | - | 2 - 3 | 659 | 74 | ||
| AK150517.1 | - | 2 - 3 | 679 | 74 | ||
| AK154955.1 | - | 2 - 3 | 665 | 74 | ||
| BC006957.1 | - | 2 - 3 | 549 | 74 | ||
| BC034829.1 | - | 2 - 3 | 669 | 4482756 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27798976 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27798979 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27798978 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27798977 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27798981 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27798980 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2009477 | BC006957.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2009477 | AK154955.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2009477 | AK147956.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2009477 | AK150517.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2009477 | AK144677.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 535371 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 420086 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 143606 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 756674 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 179409 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 397145 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 326410 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 431526 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 61801 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P100669 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P150968 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2009477 | BC006957.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2009477 | AK154955.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2009477 | AK147956.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2009477 | AK150517.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2009477 | AK144677.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27798982 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27798985 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27798965 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27798963 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27798967 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27798996 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27798978 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27798981 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27798986 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27798987 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27798983 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27798992 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27798988 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27798960 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27798974 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27798962 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27798971 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27798994 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27798990 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P100669 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07342793 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P150968 | - | - |
| Whole Mouse Genome G4122A | A_51_P392627 | BC034829.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P392627 | BC006957.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P392627 | AK154955.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P392627 | AK147956.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P392627 | AK150517.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P265762 | BC034829.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P265762 | BC006957.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P265762 | AK154955.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P265762 | AK147956.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P265762 | AK150517.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P100669 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P150968 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09303518 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27798980 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27798981 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27798978 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27798979 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27798977 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27798976 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27799077 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27799006 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27799005 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27798955 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27798978 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27799045 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27799008 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27799044 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27799063 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27799053 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27799034 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27799073 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27799075 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27799043 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27798985 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27799055 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27799066 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27798994 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27798988 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27799039 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27799061 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27798999 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27799052 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27799047 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27799010 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27799059 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27799024 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27799016 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27798966 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27799060 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27799071 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27799054 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27799080 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27799070 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27799037 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27799035 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27799068 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27799019 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27798971 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27799012 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27799046 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27798972 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27799041 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27798991 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27799078 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27799001 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27799021 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27798962 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27799051 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27798982 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27799017 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27798965 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27799004 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09702045 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27798971 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27799010 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09473544 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27799005 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27798985 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09699029 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27799026 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27798994 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09192191 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09143727 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09083397 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09192189 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09190002 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27799001 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27799012 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27798988 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27799008 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27798957 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27799021 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27798991 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27799024 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27799004 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08983729 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27798960 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09303518 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27798983 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09702043 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27798968 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27798955 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27798959 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27798978 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27799019 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27799016 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10375952 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10375953 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10375954 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10375955 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1427108_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1429921_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10375951 | - | - |
| Mouse Genome 430 2.0 Array | 1427108_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1429921_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1429921_at | - | - |
| Mouse Genome 430 2.0 Array | 1427108_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 213673_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 213673_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100004655.mm.1 | NM_153117.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100004655.mm.1 | NM_153117.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100004655.mm.1 | TC1100000593.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100004661.mm.1 | NM_153117.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100004661.mm.1 | NM_153117.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100004661.mm.1 | TC1100000593.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100004654.mm.1 | NM_153117.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100004654.mm.1 | NM_153117.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100004654.mm.1 | TC1100000593.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100004657.mm.1 | NM_153117.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100004657.mm.1 | NM_153117.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100004657.mm.1 | TC1100000593.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100004660.mm.1 | NM_153117.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100004660.mm.1 | NM_153117.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100004660.mm.1 | TC1100000593.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100004658.mm.1 | NM_153117.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100004658.mm.1 | NM_153117.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100004658.mm.1 | TC1100000593.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1427108_at | - | - |
| Mouse Expression Set 430 | 1427108_PM_at | - | - |
| Mouse Expression Set 430 | 1429921_at | - | - |
| Mouse Expression Set 430 | 1429921_PM_at | - | - |
| Murine Genome U74v2 Set | 99624_PM_at | - | - |
| Murine Genome U74v2 Set | 99624_at | - | - |
| Mouse Genome 430A 2.0 Array | 1429921_at | - | - |
| Mouse Genome 430A 2.0 Array | 1429921_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427108_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427108_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1427108_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1427108_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1429921_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1429921_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 5390132 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2768859 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1253452 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2768859 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1253452 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1253452 | - | - |

9530068E07Rik
Genomic Map