Assay Details
| Gene Symbol: Sgcd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011891.4 | NP_036021.1 | 6 - 7 | 624 | 104 | |
| GenBank mRNA | AB024923.1 | - | 6 - 7 | 845 | 104 | |
| AK040974.1 | - | 6 - 7 | 626 | 104 | ||
| BC130019.1 | - | 6 - 7 | 614 | 40140624 | 104 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sgcd
Genomic Map