Assay Details
| Gene Symbol: Slu7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_148673.3 | NP_683514.2 | 14 - 15 | 1575 | 105 | |
| NM_198936.1 | NP_945174.1 | 13 - 14 | 1496 | 105 | ||
| GenBank mRNA | AK029117.1 | - | 13 - 14 | 1466 | 105 | |
| AK046262.1 | - | 13 - 14 | 1467 | 105 | ||
| AK047597.1 | - | 14 - 15 | 1563 | 105 | ||
| AK049178.1 | - | 13 - 14 | 1465 | 105 | ||
| BC013810.1 | - | 11 - 12 | 1095 | 105 | ||
| BC023057.1 | - | 3 - 4 | 350 | 105 | ||
| BC060954.1 | - | 13 - 14 | 1464 | 105 | ||
| BC082780.1 | - | 14 - 15 | 1555 | 105 | ||
| BC106099.1 | - | 13 - 14 | 1477 | 105 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slu7
Genomic Map