Assay Details
| Gene Symbol: Slu7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_148673.3 | NP_683514.2 | 15 - 16 | 1646 | 128 | |
| NM_198936.1 | NP_945174.1 | 14 - 15 | 1567 | 128 | ||
| GenBank mRNA | AK029117.1 | - | 14 - 15 | 1537 | 128 | |
| AK046262.1 | - | 14 - 15 | 1538 | 128 | ||
| AK049178.1 | - | 14 - 15 | 1536 | 128 | ||
| BC013810.1 | - | 12 - 13 | 1166 | 128 | ||
| BC023057.1 | - | 4 - 5 | 421 | 128 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slu7
Genomic Map