Assay Details
| Gene Symbol: Slu7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_148673.3 | NP_683514.2 | 7 - 8 | 816 | 79 | |
| NM_198936.1 | NP_945174.1 | 6 - 7 | 737 | 79 | ||
| GenBank mRNA | AK029117.1 | - | 6 - 7 | 707 | 79 | |
| AK046262.1 | - | 6 - 7 | 708 | 79 | ||
| AK047597.1 | - | 7 - 8 | 804 | 79 | ||
| AK049178.1 | - | 6 - 7 | 706 | 79 | ||
| BC013810.1 | - | 4 - 5 | 336 | 79 | ||
| BC025870.1 | - | 7 - 8 | 795 | 79 | ||
| BC060954.1 | - | 6 - 7 | 705 | 79 | ||
| BC082780.1 | - | 7 - 8 | 796 | 79 | ||
| BC106099.1 | - | 6 - 7 | 718 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slu7
Genomic Map