Assay Details
| Gene Symbol: Slu7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_148673.3 | NP_683514.2 | 9 - 10 | 1002 | 114 | |
| NM_198936.1 | NP_945174.1 | 8 - 9 | 923 | 114 | ||
| GenBank mRNA | AK029117.1 | - | 8 - 9 | 893 | 114 | |
| AK046262.1 | - | 8 - 9 | 894 | 114 | ||
| AK047597.1 | - | 9 - 10 | 990 | 114 | ||
| AK049178.1 | - | 8 - 9 | 892 | 114 | ||
| BC013810.1 | - | 6 - 7 | 522 | 114 | ||
| BC060954.1 | - | 8 - 9 | 891 | 114 | ||
| BC082780.1 | - | 9 - 10 | 982 | 114 | ||
| BC106099.1 | - | 8 - 9 | 904 | 114 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slu7
Genomic Map