Assay Details
| Gene Symbol: Cdk18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008795.2 | NP_032821.1 | 2 - 3 | 427 | 71 | |
| GenBank mRNA | AK004998.1 | - | 2 - 3 | 371 | 71 | |
| AK171569.1 | - | 2 - 3 | 208 | 71 | ||
| BC119617.1 | - | 3 - 4 | 303 | 40061728 | 71 | |
| X69026.1 | - | 2 - 3 | 444 | 71 | ||
More Information
Set Membership: |
|

Cdk18
Genomic Map