Assay Details
| Gene Symbol: Cdk18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008795.2 | NP_032821.1 | 5 - 6 | 684 | 82 | |
| GenBank mRNA | AK004998.1 | - | 5 - 6 | 628 | 82 | |
| AK171569.1 | - | 5 - 6 | 465 | 82 | ||
| AK198726.1 | - | 2 - 3 | 83 | |||
| BC119617.1 | - | 6 - 7 | 560 | 40061728 | 82 | |
| X69026.1 | - | 5 - 6 | 701 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cdk18
Genomic Map