Assay Details
| Gene Symbol: Rnf13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF037205.1 | - | 1 - 2 | 79 | 67 | |
| AF037206.1 | - | 1 - 2 | 79 | 67 | ||
| AI882163.1 | - | 1 - 2 | 138 | 67 | ||
| AK034135.1 | - | 1 - 2 | 130 | 67 | ||
| AK036591.1 | - | 1 - 2 | 124 | 67 | ||
| AK050095.1 | - | 1 - 2 | 92 | 67 | ||
| AK082296.1 | - | 1 - 2 | 120 | 67 | ||
| AK139447.1 | - | 1 - 2 | 85 | 67 | ||
| AK140682.1 | - | 1 - 2 | 134 | 67 | ||
| AK158046.1 | - | 1 - 2 | 120 | 67 | ||
| AK167278.1 | - | 1 - 2 | 84 | 67 | ||
| BC058182.1 | - | 1 - 2 | 87 | 5324040 | 67 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF037205.1, AF037206.1, AK036591.1, AK082296.1, AK139447.1, AK158046.1, AK167278.1, BC058182.1 transcripts.
Set Membership: |
|

Rnf13
Genomic Map