Assay Details
| Gene Symbol: Rnf13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF037205.1 | - | 5 - 6 | 489 | 105 | |
| AF037206.1 | - | 5 - 6 | 489 | 105 | ||
| AK036591.1 | - | 5 - 6 | 534 | 105 | ||
| AK082296.1 | - | 5 - 6 | 530 | 105 | ||
| AK139447.1 | - | 5 - 6 | 495 | 105 | ||
| AK140682.1 | - | 5 - 6 | 544 | 105 | ||
| AK158046.1 | - | 5 - 6 | 530 | 105 | ||
| AK167278.1 | - | 5 - 6 | 494 | 105 | ||
| BC058182.1 | - | 5 - 6 | 497 | 5324040 | 105 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rnf13
Genomic Map