Assay Details
Gene Symbol: Colec11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027866.1 | NP_082142.1 | 6 - 7 | 91 | ||
GenBank mRNA | AK003121.1 | - | 6 - 7 | 91 | ||
BC104380.1 | - | 6 - 7 | 481 | 40050136 | 91 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
