| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1100004795.mm.1 |
NM_001252332.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100004795.mm.1 |
NM_001252332.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100004795.mm.1 |
NM_001252333.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100004795.mm.1 |
NM_001252333.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100004795.mm.1 |
NM_033144.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100004795.mm.1 |
NM_033144.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100004795.mm.1 |
TC1100000608.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100004796.mm.1 |
NM_001252332.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100004796.mm.1 |
NM_001252332.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100004796.mm.1 |
NM_001252333.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100004796.mm.1 |
NM_001252333.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100004796.mm.1 |
NM_033144.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100004796.mm.1 |
NM_033144.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100004796.mm.1 |
TC1100000608.mm.1 |
MTA TC |