Assay Details
| Gene Symbol: Sept8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252333.1 | NP_001239262.1 | 3 - 4 | 618 | 59 | |
| GenBank mRNA | AK144178.1 | - | 3 - 4 | 460 | 59 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sept8
Genomic Map