Assay Details
| Gene Symbol: Usf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF479773.1 | - | 3 - 4 | 219 | 56 | |
| AK142437.1 | - | 5 - 6 | 472 | 56 | ||
| BC049784.1 | - | 5 - 6 | 415 | 6506829 | 56 | |
| DV653810.1 | - | 5 - 6 | 346 | 56 | ||
More Information
Set Membership: |
|

Usf1
Genomic Map