Assay Details
| Gene Symbol: Rgs8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026380.3 | NP_080656.2 | 2 - 3 | 403 | 62 | |
| GenBank mRNA | AK044337.1 | - | 2 - 3 | 277 | 62 | |
| AK142301.1 | - | 2 - 3 | 272 | 62 | ||
| BC065056.1 | - | 2 - 3 | 336 | 5705033 | 62 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK044337.1, AK142301.1, BC065056.1, NM_026380.3 transcripts.
Set Membership: |
|

Rgs8
Genomic Map