| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0100011414.mm.1 |
NM_026380.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011414.mm.1 |
NM_026380.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011414.mm.1 |
TC0100001379.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100011417.mm.1 |
NM_026380.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011417.mm.1 |
NM_026380.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011417.mm.1 |
TC0100001379.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100011421.mm.1 |
NM_026380.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011421.mm.1 |
NM_026380.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011421.mm.1 |
TC0100001379.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100011415.mm.1 |
NM_026380.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011415.mm.1 |
NM_026380.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011415.mm.1 |
TC0100001379.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100011422.mm.1 |
NM_026380.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011422.mm.1 |
NM_026380.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011422.mm.1 |
TC0100001379.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100011423.mm.1 |
NM_026380.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011423.mm.1 |
NM_026380.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011423.mm.1 |
TC0100001379.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100011416.mm.1 |
NM_026380.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011416.mm.1 |
NM_026380.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011416.mm.1 |
TC0100001379.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100011420.mm.1 |
NM_026380.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011420.mm.1 |
NM_026380.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011420.mm.1 |
TC0100001379.mm.1 |
MTA TC |