Assay Details
| Gene Symbol: Rnpep | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159624.1 | NP_001153096.1 | 6 - 7 | 1272 | 148 | |
| NM_145417.3 | NP_663392.2 | 7 - 8 | 1389 | 148 | ||
| GenBank mRNA | AK031138.1 | - | 6 - 7 | 1212 | 148 | |
| AK145466.1 | - | 7 - 8 | 1393 | 149 | ||
| AK159195.1 | - | 7 - 8 | 1332 | 148 | ||
| AK159445.1 | - | 7 - 8 | 1321 | 148 | ||
| AK159625.1 | - | 7 - 8 | 1332 | 148 | ||
| BC010520.1 | - | 7 - 8 | 1350 | 148 | ||
| BC019200.1 | - | 7 - 8 | 1360 | 5041005 | 148 | |
| Gene Symbol: Dnase1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK157889.1 | - | 148 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Rnpep
Genomic Map