Assay Details
| Gene Symbol: Cfap126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081275.1 | NP_001074744.1 | 2 - 3 | 304 | 66 | |
| GenBank mRNA | AK005804.1 | - | 2 - 3 | 305 | 66 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK005804.1, NM_001081275.1 transcripts.
Set Membership: |
|

Cfap126
Genomic Map