Assay Details
| Gene Symbol: Cfap126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081275.1 | NP_001074744.1 | 3 - 4 | 368 | 83 | |
| GenBank mRNA | BC061017.1 | - | 2 - 3 | 94 | 6774925 | 83 |
More Information
Updated Mapping Information
Set Membership: |
|

Cfap126
Genomic Map