Assay Details
| Gene Symbol: 4930447C04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029444.2 | NP_083720.1 | 10 - 11 | 943 | 99 | |
| GenBank mRNA | AK015397.1 | - | 10 - 11 | 943 | 99 | |
| BC137875.1 | - | 11 - 12 | 1021 | 8860894 | 99 | |
More Information
Updated Mapping Information
Set Membership: |
|

4930447C04Rik
Genomic Map