Assay Details
| Gene Symbol: Mpzl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001880.2 | NP_001001880.1 | 4 - 5 | 754 | 81 | |
| GenBank mRNA | AK076882.1 | - | 4 - 5 | 587 | 81 | |
| AK189245.1 | - | 3 - 4 | 417 | 81 | ||
| BC072563.1 | - | 4 - 5 | 749 | 6808882 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mpzl1
Genomic Map