| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0100022778.mm.1 |
NM_001163425.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100022778.mm.1 |
NM_001163425.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100022778.mm.1 |
NR_028108.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100022778.mm.1 |
NR_028108.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100022778.mm.1 |
TC0100002816.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100022774.mm.1 |
NM_001163425.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100022774.mm.1 |
NM_001163425.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100022774.mm.1 |
NR_028108.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100022774.mm.1 |
NR_028108.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100022774.mm.1 |
TC0100002816.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100022779.mm.1 |
NM_001163425.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100022779.mm.1 |
NM_001163425.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100022779.mm.1 |
NR_028108.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100022779.mm.1 |
NR_028108.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100022779.mm.1 |
TC0100002816.mm.1 |
MTA TC |