Assay Details
| Gene Symbol: Mettl11b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001143956.1 | NP_001137428.1 | 2 - 3 | 505 | 69 | |
| GenBank mRNA | BC157906.1 | - | 2 - 3 | 505 | 9088108 | 69 |
| BC158115.1 | - | 2 - 3 | 482 | 9088188 | 69 | |
| BC172048.1 | - | 2 - 3 | 505 | 9054334 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mettl11b
Genomic Map