Assay Details
| Gene Symbol: Wdr41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK082348.1 | - | 9 - 10 | 894 | 77 | |
| BC145143.1 | - | 9 - 10 | 795 | 77 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wdr41| Gene Symbol: Wdr41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK082348.1 | - | 9 - 10 | 894 | 77 | |
| BC145143.1 | - | 9 - 10 | 795 | 77 | ||
Set Membership: |
|