Assay Details
| Gene Symbol: Sept8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252332.1 | NP_001239261.1 | 3 - 4 | 626 | 64 | |
| NM_033144.2 | NP_149156.1 | 3 - 4 | 626 | 64 | ||
| GenBank mRNA | AB093215.1 | - | 3 - 4 | 476 | 64 | |
| AF179996.1 | - | 2 - 3 | 322 | 64 | ||
| BC037707.1 | - | 3 - 4 | 414 | 64 | ||
| BC049819.1 | - | 3 - 4 | 441 | 64 | ||
| BC059248.1 | - | 3 - 4 | 562 | 6400429 | 64 | |
More Information
Set Membership: |
|

Sept8
Genomic Map