Assay Details
| Gene Symbol: Ankrd34b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175455.4 | NP_780664.2 | 2 - 3 | 378 | 69 | |
| GenBank mRNA | AK078203.1 | - | 2 - 3 | 452 | 69 | |
| AK090387.1 | - | 2 - 3 | 379 | 69 | ||
| AK138465.1 | - | 1 - 2 | 564 | 69 | ||
| BC096609.1 | - | 2 - 3 | 254 | 69 | ||
| BC119312.1 | - | 2 - 3 | 240 | 8734061 | 69 | |
| BC119314.1 | - | 2 - 3 | 240 | 8734063 | 69 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK078203.1, AK138465.1, BC096609.1, BC119312.1, BC119314.1, NM_175455.4 transcripts.
Set Membership: |
|

Ankrd34b
Genomic Map