Assay Details
| Gene Symbol: Acmsd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033041.2 | NP_001028213.1 | 6 - 7 | 653 | 76 | |
| GenBank mRNA | AK143962.1 | - | 6 - 7 | 655 | 76 | |
| BC139281.1 | - | 6 - 7 | 604 | 8862303 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Acmsd
Genomic Map