Assay Details
| Gene Symbol: Snx13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014973.2 | NP_001014973.2 | 24 - 25 | 2727 | 62 | |
| GenBank mRNA | AK122352.1 | - | 13 - 14 | 1376 | 62 | |
| AK133353.1 | - | 21 - 22 | 2267 | 62 | ||
| AK164161.1 | - | 24 - 25 | 2652 | 62 | ||
| BC056394.1 | - | 24 - 25 | 2699 | 6852346 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Snx13
Genomic Map