Assay Details
| Gene Symbol: Rnf13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF037205.1 | - | 6 - 7 | 591 | 111 | |
| AF037206.1 | - | 6 - 7 | 591 | 111 | ||
| AK034135.1 | - | 6 - 7 | 652 | 111 | ||
| AK036591.1 | - | 6 - 7 | 636 | 111 | ||
| AK050095.1 | - | 6 - 7 | 1336 | 111 | ||
| AK082296.1 | - | 6 - 7 | 632 | 111 | ||
| AK139447.1 | - | 6 - 7 | 597 | 111 | ||
| AK140682.1 | - | 6 - 7 | 646 | 111 | ||
| AK158046.1 | - | 6 - 7 | 632 | 111 | ||
| AK167278.1 | - | 6 - 7 | 596 | 111 | ||
| BC058182.1 | - | 6 - 7 | 599 | 5324040 | 111 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rnf13
Genomic Map