Assay Details
Gene Symbol: Slc36a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153139.4 | NP_694779.3 | 3 - 4 | 387 | 111 | |
GenBank mRNA | AF453743.1 | - | 2 - 3 | 373 | 111 | |
AY211262.1 | - | 3 - 4 | 357 | 111 | ||
BC138556.1 | - | 3 - 4 | 351 | 8861576 | 111 | |
BC138558.1 | - | 3 - 4 | 351 | 8861578 | 111 | |
CJ105223.1 | - | 388 | 111 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
