Assay Details
Gene Symbol: Slc36a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153139.4 | NP_694779.3 | 5 - 6 | 573 | 66 | |
GenBank mRNA | AF453743.1 | - | 4 - 5 | 559 | 66 | |
AY211262.1 | - | 5 - 6 | 543 | 66 | ||
BC138556.1 | - | 5 - 6 | 537 | 8861576 | 66 | |
BC138558.1 | - | 5 - 6 | 537 | 8861578 | 66 | |
CJ105223.1 | - | 574 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
