Assay Details
| Gene Symbol: Trpc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC108988.2 | - | 4 - 5 | 1302 | 77 | |
| BC108989.2 | - | 4 - 5 | 1303 | 40047766 | 77 | |
| FJ207475.1 | - | 4 - 5 | 1303 | 77 | ||
| FJ207476.1 | - | 4 - 5 | 1303 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Trpc3
Genomic Map