Assay Details
| Gene Symbol: Trpc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019510.2 | NP_062383.2 | 8 - 9 | 2409 | 93 | |
| GenBank mRNA | AF190645.1 | - | 7 - 8 | 1999 | 93 | |
| AK032317.1 | - | 8 - 9 | 2410 | 93 | ||
| AK080619.1 | - | 8 - 9 | 2397 | 93 | ||
| AK163714.1 | - | 2 - 3 | 399 | 93 | ||
| BC108988.2 | - | 7 - 8 | 1998 | 93 | ||
| BC108989.2 | - | 7 - 8 | 1999 | 40047766 | 93 | |
| FJ207475.1 | - | 7 - 8 | 1999 | 93 | ||
| FJ207476.1 | - | 7 - 8 | 1999 | 93 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Trpc3
Genomic Map