Assay Details
| Gene Symbol: Arhgap5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009706.2 | NP_033836.2 | 1 - 2 | 3890 | 107 | |
| GenBank mRNA | BC150823.1 | - | 1 - 2 | 3860 | 9087734 | 107 |
| CF742308.1 | - | 1 - 2 | 156 | 107 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Arhgap5
Genomic Map