Assay Details
| Gene Symbol: Shcbp1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033162.2 | NP_001028334.1 | 7 - 8 | 1311 | 83 | |
| GenBank mRNA | AK005886.1 | - | 7 - 8 | 1308 | 83 | |
| AK161274.1 | - | 7 - 8 | 1312 | 83 | ||
| BC113157.1 | - | 7 - 8 | 1358 | 40058375 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|

Shcbp1l
Genomic Map