Assay Details
| Gene Symbol: Shcbp1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033162.2 | NP_001028334.1 | 9 - 10 | 1683 | 115 | |
| GenBank mRNA | AK005886.1 | - | 9 - 10 | 1680 | 115 | |
| AK161274.1 | - | 9 - 10 | 1684 | 115 | ||
| BC113157.1 | - | 9 - 10 | 1730 | 40058375 | 115 | |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20726626 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20726627 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20726623 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P266077 | BC113157.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P266077 | AK005886.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P266077 | AK161274.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P266077 | NM_001033162.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P266077 | BC113157.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P266077 | AK005886.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P266077 | AK161274.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P266077 | NM_001033162.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 862969 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 117182 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 587648 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 543918 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 267179 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 248320 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 453328 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 179207 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 566178 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 779575 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 193638 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 922393 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 692108 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 175599 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 808034 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 29353 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 51180 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 137858 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 85012 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P139862 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P266077 | BC113157.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P266077 | AK005886.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P266077 | AK161274.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P266077 | NM_001033162.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P266077 | BC113157.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P266077 | AK005886.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P266077 | AK161274.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P266077 | NM_001033162.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20726611 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20726624 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20726630 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20726613 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20726621 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20726635 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20726632 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20726638 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20726637 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20726619 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20726627 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20726617 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20726615 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20726628 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20726625 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20726616 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04311546 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00219615 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P139862 | - | - |
| Whole Mouse Genome G4122A | A_51_P266077 | BC113157.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P266077 | AK005886.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P266077 | AK161274.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P266077 | NM_001033162.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P266077 | BC113157.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P266077 | AK005886.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P266077 | AK161274.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P266077 | NM_001033162.2 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P04311546 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P139862 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20726626 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20726623 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20726627 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20726758 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20726764 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20726738 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20726798 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20726678 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20726748 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20726663 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20726628 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20726725 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20726731 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20726739 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20726823 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20726789 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20726712 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20726635 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20726806 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20726819 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20726641 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20726654 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20726619 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20726705 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20726813 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20726785 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20726676 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20726675 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20726659 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20726643 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20726689 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20726697 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20726673 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20726700 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20726616 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20726817 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20726681 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20726735 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20726693 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20726698 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20726633 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20726638 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20726706 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20726791 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20726624 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20726801 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20726646 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20726746 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20726753 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20726810 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20726763 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20726720 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20726703 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20726825 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20726683 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20726821 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20726666 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20726750 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20726709 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20726625 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20726687 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20726796 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20726702 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20726615 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20726794 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20726727 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20726766 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20726648 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20726692 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20726793 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20726729 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20726804 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20726656 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20726685 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20726800 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20726787 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20726809 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20726815 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20726695 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20726617 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20726650 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20726742 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20726816 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20726711 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20726797 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20726671 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20726728 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20726710 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20726761 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20726707 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00847011 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20726617 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20726654 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20726613 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00768099 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20726616 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00413974 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00625185 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00872588 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20726600 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20726650 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00413976 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20726633 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00625190 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00944652 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00844829 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20726641 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00978786 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20726638 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20726631 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20726658 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20726637 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20726632 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20726612 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20726621 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20726611 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00413979 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20726646 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20726652 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20726648 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00847012 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20726630 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00759050 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10350720 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10350713 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10350715 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10350712 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10350722 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10350717 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10350716 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10350721 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10350719 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10350714 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10350718 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1429324_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10350710 | - | - |
| Mouse Genome 430 2.0 Array | 1429324_at | - | - |
| Mouse Genome 430 2.0 Array | 1429324_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 71836_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 71836_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0100011393.mm.1 | NM_001033162.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100011393.mm.1 | NM_001033162.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100011393.mm.1 | TC0100001377.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100011392.mm.1 | NM_001033162.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100011392.mm.1 | NM_001033162.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100011392.mm.1 | TC0100001377.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100011394.mm.1 | NM_001033162.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100011394.mm.1 | NM_001033162.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100011394.mm.1 | TC0100001377.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1429324_at | - | - |
| Mouse Expression Set 430 | 1429324_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429324_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429324_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 342444 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1218141 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1218141 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1218141 | - | - |

Shcbp1l
Genomic Map