| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1200011139.mm.1 |
NM_153198.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200011139.mm.1 |
NM_153198.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200011139.mm.1 |
NM_177993.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200011139.mm.1 |
NM_177993.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200011139.mm.1 |
TC1200001601.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200011148.mm.1 |
NM_153198.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200011148.mm.1 |
NM_153198.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200011148.mm.1 |
NM_177993.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200011148.mm.1 |
NM_177993.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200011148.mm.1 |
TC1200001601.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200011137.mm.1 |
NM_153198.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200011137.mm.1 |
NM_153198.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200011137.mm.1 |
NM_177993.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200011137.mm.1 |
NM_177993.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200011137.mm.1 |
TC1200001601.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200011138.mm.1 |
NM_153198.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200011138.mm.1 |
NM_153198.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200011138.mm.1 |
NM_177993.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200011138.mm.1 |
NM_177993.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200011138.mm.1 |
TC1200001601.mm.1 |
MTA TC |