Assay Details
| Gene Symbol: Arl8a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026823.2 | NP_081099.1 | 6 - 7 | 657 | 91 | |
| GenBank mRNA | AB185207.1 | - | 6 - 7 | 513 | 91 | |
| AK004080.1 | - | 3 - 4 | 185 | 91 | ||
| BC004035.1 | - | 5 - 6 | 388 | 91 | ||
| BC018479.1 | - | 6 - 7 | 657 | 3661329 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20637765 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20637766 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20637770 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20637774 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20637773 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20637771 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20637767 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20637768 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20637772 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20637769 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P224593 | BC018479.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P224593 | NM_026823.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P224593 | AK004080.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P224593 | BC004035.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P224593 | BC018479.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P224593 | NM_026823.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P224593 | AK004080.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P224593 | BC004035.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 400043 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 34606 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 382716 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 190343 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 10348 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 641423 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P138150 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P224593 | BC018479.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P224593 | NM_026823.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P224593 | AK004080.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P224593 | BC004035.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P224593 | BC018479.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P224593 | NM_026823.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P224593 | AK004080.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P224593 | BC004035.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P224593 | BC018479.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P224593 | NM_026823.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P224593 | AK004080.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P224593 | BC004035.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P224593 | BC018479.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P224593 | NM_026823.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P224593 | AK004080.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P224593 | BC004035.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20637773 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20637781 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20637783 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20637766 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20637776 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20637758 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20637775 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20637788 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20637757 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20637784 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20637786 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20637765 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20637763 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20637769 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20637780 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20637760 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20637779 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20637771 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20637777 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04273888 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P138150 | - | - |
| Whole Mouse Genome G4122A | A_51_P224593 | BC018479.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P224593 | NM_026823.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P224593 | AK004080.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P224593 | BC004035.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P224593 | BC018479.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P224593 | NM_026823.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P224593 | AK004080.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P224593 | BC004035.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P137030 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P138150 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00563055 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00563056 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00563051 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00563048 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20637769 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20637765 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20637771 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20637773 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20637772 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20637766 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20637774 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20637770 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20637767 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20637768 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20637825 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20637769 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20637763 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20637827 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20637853 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20637850 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20637873 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20637881 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20637801 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20637788 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20637862 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20637805 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20637838 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20637883 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20637848 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20637870 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20637794 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20637797 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20637766 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20637784 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20637765 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20637852 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20637836 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20637832 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20637711 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20637773 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20637857 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20637849 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20637779 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20637864 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20637851 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20637841 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20637885 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20637809 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20637783 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20637892 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20637818 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20637824 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20637707 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20637807 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20637831 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20637767 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20637760 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20637776 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20637708 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20637791 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20637842 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20637872 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20637856 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20637759 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20637875 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20637811 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20637799 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20637855 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20637781 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20637877 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20637790 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20637834 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20637806 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20637868 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20637714 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20637867 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20637771 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20637777 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20637719 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20637812 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20637845 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20637859 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20637816 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20637803 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20637821 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20637829 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20637786 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20637826 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20637705 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20637823 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20637807 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20637794 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00327253 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20637799 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01067466 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01096396 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20637797 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20637790 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01096395 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01067473 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01067468 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20637769 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20637801 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00954799 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01067478 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20637783 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20637803 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01011466 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00790043 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20637846 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20637805 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20637811 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20637809 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20637776 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20637813 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20637788 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20637793 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00563048 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20637791 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01096400 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20637766 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20637786 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00563055 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00137479 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01096397 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01096401 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00563056 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10350118 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10350116 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10350114 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10350121 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10350120 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10350117 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10350122 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10350119 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10350115 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1424640_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10350113 | - | - |
| Mouse Genome 430 2.0 Array | 1424640_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1424640_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68724_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68724_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0100010246.mm.1 | NM_026823.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100010246.mm.1 | NM_026823.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100010246.mm.1 | TC0100001245.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100010245.mm.1 | NM_026823.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100010245.mm.1 | NM_026823.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100010245.mm.1 | TC0100001245.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1424640_at | - | - |
| Mouse Expression Set 430 | 1424640_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424640_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424640_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424640_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424640_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 470494 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 4810162 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4810162 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004810162 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2680781 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2680781 | - | - |
| mouse-6 v1.1 (Illumina) | scl17443.8_550-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4810162 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl17443.8_550-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4810162 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2680781 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2680781 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl17443.8_550-S | - | - |

Arl8a
Genomic Map