Assay Details
| Gene Symbol: Jade2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199299.3 | NP_955003.2 | 3 - 4 | 324 | 61 | |
| GenBank mRNA | AK004823.1 | - | 3 - 4 | 496 | 61 | |
| AK129097.1 | - | 3 - 4 | 324 | 61 | ||
| AK147453.1 | - | 4 - 5 | 328 | 61 | ||
| BC117856.1 | - | 2 - 3 | 153 | 40094330 | 61 | |
| BC117857.1 | - | 2 - 3 | 153 | 40094331 | 61 | |
More Information
Set Membership: |
|

Jade2
Genomic Map