Assay Details
| Gene Symbol: Jade2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199299.3 | NP_955003.2 | 6 - 7 | 859 | 68 | |
| GenBank mRNA | AK004823.1 | - | 6 - 7 | 1031 | 68 | |
| AK129097.1 | - | 6 - 7 | 859 | 68 | ||
| AK147453.1 | - | 7 - 8 | 863 | 68 | ||
| BC117856.1 | - | 5 - 6 | 688 | 40094330 | 68 | |
| BC117857.1 | - | 5 - 6 | 688 | 40094331 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Jade2
Genomic Map