Assay Details
| Gene Symbol: Jade2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199299.3 | NP_955003.2 | 8 - 9 | 1141 | 120 | |
| GenBank mRNA | AK004823.1 | - | 8 - 9 | 1313 | 120 | |
| AK129097.1 | - | 8 - 9 | 1141 | 120 | ||
| AK147453.1 | - | 9 - 10 | 1145 | 120 | ||
| BC117856.1 | - | 7 - 8 | 970 | 40094330 | 120 | |
| BC117857.1 | - | 7 - 8 | 970 | 40094331 | 120 | |
More Information
Updated Mapping Information
Set Membership: |
|

Jade2
Genomic Map