Assay Details
| Gene Symbol: Jade2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199299.3 | NP_955003.2 | 9 - 10 | 1603 | 78 | |
| GenBank mRNA | AK004823.1 | - | 9 - 10 | 1782 | 78 | |
| AK129097.1 | - | 9 - 10 | 1603 | 78 | ||
| AK147453.1 | - | 10 - 11 | 1607 | 78 | ||
| BC117856.1 | - | 8 - 9 | 1432 | 40094330 | 78 | |
| BC117857.1 | - | 8 - 9 | 1432 | 40094331 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Jade2
Genomic Map