Assay Details
| Gene Symbol: Etnk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175443.5 | NP_780652.2 | 6 - 7 | 1204 | 61 | |
| GenBank mRNA | AK077179.1 | - | 6 - 7 | 943 | 61 | |
| AK087818.1 | - | 5 - 6 | 1002 | 61 | ||
| BC152310.1 | - | 6 - 7 | 1142 | 40089976 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|

Etnk2
Genomic Map