Assay Details
| Gene Symbol: Etnk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175443.5 | NP_780652.2 | 7 - 8 | 1281 | 104 | |
| GenBank mRNA | AK077179.1 | - | 7 - 8 | 1020 | 104 | |
| AK087818.1 | - | 6 - 7 | 1079 | 104 | ||
| BC152310.1 | - | 7 - 8 | 1219 | 40089976 | 104 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
in utero embryonic development
placenta development
lipid metabolic process
phosphatidylethanolamine biosynthetic process
phospholipid biosynthetic process
post-embryonic development
phosphorylation
multicellular organism growth
nucleotide binding
choline kinase activity
ethanolamine kinase activity
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
in utero embryonic development
placenta development
lipid metabolic process
phosphatidylethanolamine biosynthetic process
phospholipid biosynthetic process
post-embryonic development
phosphorylation
multicellular organism growth
nucleotide binding
choline kinase activity
ethanolamine kinase activity
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P238183 | AU256212.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P238183 | AK087818.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P238183 | AK077179.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P238183 | NM_175443.5 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P238183 | AU256212.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P238183 | AK087818.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P238183 | AK077179.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P238183 | NM_175443.5 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1979019 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 425746 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 790321 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 37326 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 232243 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 223506 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 745154 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 109512 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 434043 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 940185 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 335187 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 54381 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 583350 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 139529 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P120378 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P127361 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2034285 | AK031665.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P238183 | AU256212.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P238183 | AK087818.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P238183 | AK077179.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P238183 | NM_175443.5 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P238183 | AU256212.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P238183 | AK087818.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P238183 | AK077179.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P238183 | NM_175443.5 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1979019 | - | - |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P165824 | AK031665.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P238183 | AU256212.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P238183 | AK087818.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P238183 | AK077179.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P238183 | NM_175443.5 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P238183 | AU256212.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P238183 | AK087818.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P238183 | AK077179.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P238183 | NM_175443.5 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P165824 | AK031665.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P101447 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01105852 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01105849 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00618520 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10349940 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10349938 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10349933 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10349936 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10349935 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10349941 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10349934 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10349942 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10349939 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1437040_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10349932 | - | - |
| Mouse Genome 430 2.0 Array | 1437040_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1437040_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0100009919.mm.1 | NM_175443.5.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100009919.mm.1 | NM_175443.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100009919.mm.1 | TC0100001207.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100009920.mm.1 | NM_175443.5.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100009920.mm.1 | NM_175443.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100009920.mm.1 | TC0100001207.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1437040_PM_at | - | - |
| Mouse Expression Set 430 | 1437040_at | - | - |
| Murine Genome U74v2 Set | 166043_PM_at | - | - |
| Murine Genome U74v2 Set | 166043_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1437040_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1437040_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 105910039 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4540373 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004540373 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2612868 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2476940 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2612868 | - | - |
| mouse-6 v1.1 (Illumina) | scl00214253.2_23-S | - | - |
| mouse-6 v1.1 (Illumina) | scl17463.8_535-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4540373 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 105910039 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl00214253.2_23-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4540373 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 105910039 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2612868 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2612868 | - | - |

Etnk2
Genomic Map