| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1200012570.mm.1 |
NM_001193266.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200012570.mm.1 |
NM_001193266.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200012570.mm.1 |
NM_207010.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200012570.mm.1 |
NM_207010.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200012570.mm.1 |
TC1200001843.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200012575.mm.1 |
NM_001193266.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200012575.mm.1 |
NM_001193266.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200012575.mm.1 |
NM_207010.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200012575.mm.1 |
NM_207010.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200012575.mm.1 |
TC1200001843.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200012582.mm.1 |
NM_001193266.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200012582.mm.1 |
NM_001193266.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200012582.mm.1 |
NM_207010.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200012582.mm.1 |
NM_207010.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200012582.mm.1 |
TC1200001843.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200012580.mm.1 |
NM_001193266.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200012580.mm.1 |
NM_001193266.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200012580.mm.1 |
NM_207010.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200012580.mm.1 |
NM_207010.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200012580.mm.1 |
TC1200001843.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200012567.mm.1 |
NM_001193266.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200012567.mm.1 |
NM_001193266.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200012567.mm.1 |
NM_207010.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200012567.mm.1 |
NM_207010.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200012567.mm.1 |
TC1200001843.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200012569.mm.1 |
NM_001193266.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200012569.mm.1 |
NM_001193266.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200012569.mm.1 |
NM_207010.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200012569.mm.1 |
NM_207010.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200012569.mm.1 |
TC1200001843.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200012571.mm.1 |
NM_001193266.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200012571.mm.1 |
NM_001193266.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200012571.mm.1 |
NM_207010.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200012571.mm.1 |
NM_207010.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200012571.mm.1 |
TC1200001843.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200012584.mm.1 |
NM_001193266.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200012584.mm.1 |
NM_001193266.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200012584.mm.1 |
NM_207010.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200012584.mm.1 |
NM_207010.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200012584.mm.1 |
TC1200001843.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200012568.mm.1 |
NM_001193266.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200012568.mm.1 |
NM_001193266.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200012568.mm.1 |
NM_207010.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200012568.mm.1 |
NM_207010.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200012568.mm.1 |
TC1200001843.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200012572.mm.1 |
NM_001193266.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200012572.mm.1 |
NM_001193266.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200012572.mm.1 |
NM_207010.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200012572.mm.1 |
NM_207010.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200012572.mm.1 |
TC1200001843.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200012583.mm.1 |
NM_001193266.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200012583.mm.1 |
NM_001193266.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200012583.mm.1 |
NM_207010.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200012583.mm.1 |
NM_207010.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200012583.mm.1 |
TC1200001843.mm.1 |
MTA TC |