Assay Details
| Gene Symbol: Mdga2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193266.1 | NP_001180195.2 | 7 - 8 | 2819 | 113 | |
| NM_207010.2 | NP_996893.3 | 7 - 8 | 2819 | 113 | ||
| GenBank mRNA | AK137667.1 | - | 7 - 8 | 2820 | 113 | |
| AY371925.1 | - | 7 - 8 | 1473 | 113 | ||
| BC137901.1 | - | 7 - 8 | 1472 | 8860920 | 113 | |
| BC137902.1 | - | 7 - 8 | 1472 | 8860921 | 113 | |
| BC145021.1 | - | 7 - 8 | 1467 | 113 | ||
| BC171938.1 | - | 7 - 8 | 1472 | 9054224 | 113 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mdga2
Genomic Map