Assay Details
| Gene Symbol: Mdga2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207010.2 | NP_996893.3 | 11 - 12 | 3717 | 137 | |
| GenBank mRNA | AY371925.1 | - | 11 - 12 | 2371 | 137 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mdga2
Genomic Map