Assay Details
| Gene Symbol: Mdga2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193266.1 | NP_001180195.2 | 13 - 14 | 3884 | 67 | |
| NM_207010.2 | NP_996893.3 | 12 - 13 | 3863 | 67 | ||
| GenBank mRNA | AK137667.1 | - | 13 - 14 | 3885 | 67 | |
| AY371925.1 | - | 12 - 13 | 2517 | 67 | ||
| BC137901.1 | - | 13 - 14 | 2537 | 8860920 | 67 | |
| BC137902.1 | - | 13 - 14 | 2537 | 8860921 | 67 | |
| BC145021.1 | - | 13 - 14 | 2532 | 67 | ||
| BC171938.1 | - | 13 - 14 | 2537 | 9054224 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mdga2
Genomic Map