Assay Details
| Gene Symbol: Mdga2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193266.1 | NP_001180195.2 | 14 - 15 | 4040 | 133 | |
| NM_207010.2 | NP_996893.3 | 13 - 14 | 4019 | 133 | ||
| GenBank mRNA | AK137667.1 | - | 14 - 15 | 4041 | 133 | |
| AY371925.1 | - | 13 - 14 | 2673 | 133 | ||
| BC137901.1 | - | 14 - 15 | 2693 | 8860920 | 133 | |
| BC137902.1 | - | 14 - 15 | 2693 | 8860921 | 133 | |
| BC145021.1 | - | 14 - 15 | 2688 | 133 | ||
| BC171938.1 | - | 14 - 15 | 2693 | 9054224 | 133 | |
More Information
Set Membership: |
|

Mdga2
Genomic Map