Assay Details
| Gene Symbol: Hspa2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002012.1 | NP_001002012.1 | 1 - 2 | 115 | 66 | |
| GenBank mRNA | BC052350.1 | - | 1 - 2 | 115 | 6772789 | 66 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC052350.1, NM_001002012.1 transcripts.
Set Membership: |
|

Hspa2
Genomic Map